Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexane ReagentPlusR 99500ML
Catalog No. 502898242
Encompass
Hexane is an alkane and exists in 5 structural isomeric forms i.e. hexane isohexane 3-methylpentane neohexane and 2 3-dimethylbutane. It can be obtained from petroleum by distillation. It can also be produced by electrolyzing the potassium butyrate SOLUTIONon. It is widely employed organic solvent for the disSOLUTIONon and isolation of fats and oils.
Catalog No. 50-289-8242 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1393866X500ML
Hexane is an alkane and exists in 5 structural isomeric forms i.e. hexane isohexane 3-methylpentane neohexane and 2 3-dimethylbutane. It can be obtained from petroleum by distillation. It can also be produced by electrolyzing the potassium butyrate SOLUTIONon. It is widely employed organic solvent for the disSOLUTIONon and isolation of fats and oils.