Learn More
Medchemexpress LLC Hexanedioic-3,3,4,4-d4 acid, dimethyl ester | 55724-08-8 | MFCD12022191 | 99.4% | 178.22 g/mol | C8H10D4O4 | 25 MG
SDP
Catalog No. 5000303530
Encompass_Preferred
Dimethyl adipate-d4-1 is a deuterium-labeled analogue of dimethyl adipate used as an isotope-labeled analytical standard and tracer for quantitative mass spectrometry, NMR, and other analytical techniques. It provides high isotopic enrichment and chemical purity to support accurate identification and quantitation in research and quality-control workflows.
  • Deuterium-labeled internal standard for GC-MS and LC-MS
  • High isotopic enrichment for reliable quantitation
  • High chemical purity suitable for analytical use
  • Stable when stored under recommended conditions
  • Suitable as a tracer and calibration reference in analytical methods
Catalog No. 50-003-03530 Supplier Medchemexpress LLC Supplier No. 25MGHYW015301S1
Dimethyl adipate-d4-1 is a deuterium-labeled analogue of dimethyl adipate used as an isotope-labeled analytical standard and tracer for quantitative mass spectrometry, NMR, and other analytical techniques. It provides high isotopic enrichment and chemical purity to support accurate identification and quantitation in research and quality-control workflows.
  • Deuterium-labeled internal standard for GC-MS and LC-MS
  • High isotopic enrichment for reliable quantitation
  • High chemical purity suitable for analytical use
  • Stable when stored under recommended conditions
  • Suitable as a tracer and calibration reference in analytical methods