Learn More
Medchemexpress LLC Hexanedioic acid-d8 | 52089-65-3 | 99.7% | 154.19 | C6H2D8O4 | 5 MG
SDP
Catalog No. 5000299431
Encompass_Preferred
Adipic acid-d8 is a deuterium-labeled analogue of adipic acid used as a stable isotope reference standard for analytical and research applications. It is intended for use as an internal standard in mass spectrometry, method validation, and quantitative analysis where isotopic differentiation is required. The product is offered in small milligram quantities and supplied at high purity.
Catalog No. 50-002-99431 Supplier Medchemexpress LLC Supplier No. HYW017522S45MG
Adipic acid-d8 is a deuterium-labeled analogue of adipic acid used as a stable isotope reference standard for analytical and research applications. It is intended for use as an internal standard in mass spectrometry, method validation, and quantitative analysis where isotopic differentiation is required. The product is offered in small milligram quantities and supplied at high purity.