Learn More
Medchemexpress LLC Hexanedioic acid, dimethyl ester | 627-93-0 | MFCD00008469 | 99.98% | 174.19 g·mol⁻¹ | C8H14O4 | 25 G
SDP
Catalog No. 5000303476
Encompass_Preferred
Dimethyl adipate (hexanedioic acid dimethyl ester) is an aliphatic dicarboxylate ester used as a plasticizer and as an intermediate in organic synthesis. It is a colorless liquid with formula C8H14O4 and is supplied for laboratory research and analytical applications.
  • High purity (99.98%) suitable for research use.
  • Colorless liquid, easy to handle and measure.
  • Used as a plasticizer in polymer formulations.
  • Useful intermediate for organic synthesis and analytical standards.
  • Soluble in common organic solvents such as DMSO.
  • Available in multiple laboratory pack sizes for convenient use.
Catalog No. 50-003-03476 Supplier Medchemexpress LLC Supplier No. 25GHYW015301
Dimethyl adipate (hexanedioic acid dimethyl ester) is an aliphatic dicarboxylate ester used as a plasticizer and as an intermediate in organic synthesis. It is a colorless liquid with formula C8H14O4 and is supplied for laboratory research and analytical applications.
  • High purity (99.98%) suitable for research use.
  • Colorless liquid, easy to handle and measure.
  • Used as a plasticizer in polymer formulations.
  • Useful intermediate for organic synthesis and analytical standards.
  • Soluble in common organic solvents such as DMSO.
  • Available in multiple laboratory pack sizes for convenient use.