Learn More
Medchemexpress LLC Hexanedioic acid dimethyl ester-d4 | 55724-08-8 | 99.4% | 178.22 g/mol | C8H10D4O4 | 5 MG
SDP
Catalog No. 5000303737
Encompass_Preferred
Dimethyl adipate-d4 is the deuterium-labeled (d4) isotopic form of dimethyl adipate (hexanedioic acid dimethyl ester). It is supplied as a high-purity analytical standard for research and analytical workflows, commonly used as an internal standard in mass spectrometry and as a synthetic intermediate in organic synthesis.
  • Deuterium-labeled (d4) isotopic standard suitable for mass spectrometry.
  • High purity (99.4%) for reliable quantitation.
  • Molecular weight 178.22 g/mol for accurate mass calibration.
  • Available in small-scale quantities including 5 mg for trace analysis.
  • Suitable as an internal standard and analytical reference in method development.
Catalog No. 50-003-03737 Supplier Medchemexpress LLC Supplier No. HYW015301S15MG
Dimethyl adipate-d4 is the deuterium-labeled (d4) isotopic form of dimethyl adipate (hexanedioic acid dimethyl ester). It is supplied as a high-purity analytical standard for research and analytical workflows, commonly used as an internal standard in mass spectrometry and as a synthetic intermediate in organic synthesis.
  • Deuterium-labeled (d4) isotopic standard suitable for mass spectrometry.
  • High purity (99.4%) for reliable quantitation.
  • Molecular weight 178.22 g/mol for accurate mass calibration.
  • Available in small-scale quantities including 5 mg for trace analysis.
  • Suitable as an internal standard and analytical reference in method development.