Learn More
Medchemexpress LLC Hexanedioic acid dimethyl ester-d8 | 52089-64-2 | 10mg
SDP
Catalog No. 5000303607
Encompass_Preferred
Dimethyl adipate-d8 (Hexanedioic acid dimethyl ester-d8) is the deuterium labeled Dimethyl adipate (HY-W015301) Dimethyl adipate is an aliphatic dicarboxylate compound Dimethyl adipate is mainly used as a plasticizer and an intermediate in organic synthesis
Catalog No. 50-003-03607 Supplier Medchemexpress LLC Supplier No. HYW015301S210MG
Dimethyl adipate-d8 (Hexanedioic acid dimethyl ester-d8) is the deuterium labeled Dimethyl adipate (HY-W015301) Dimethyl adipate is an aliphatic dicarboxylate compound Dimethyl adipate is mainly used as a plasticizer and an intermediate in organic synthesis