Learn More
Medchemexpress LLC Hexanedioic acid, dimethyl ester-d8 | 52089-64-2 | MFCD12022192 | 98.0% | 182.24 g/mol | C8H6D8O4 | 25 MG
SDP
Catalog No. 5000304041
Encompass_Preferred
Dimethyl adipate-d8 is the deuterium-labeled analog of dimethyl adipate, provided as an analytical standard and tracer for quantitative research. It is intended for use as an internal standard in mass spectrometry and isotope-dilution assays, and as a labeled reference for method development and metabolic studies.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for isotope-dilution quantitative assays.
  • High chemical purity for reliable analytical results.
  • Stable when stored under recommended low-temperature conditions.
  • Available in multiple small sizes for analytical workflows.
Catalog No. 50-003-04041 Supplier Medchemexpress LLC Supplier No. 25MGHYW015301S2
Dimethyl adipate-d8 is the deuterium-labeled analog of dimethyl adipate, provided as an analytical standard and tracer for quantitative research. It is intended for use as an internal standard in mass spectrometry and isotope-dilution assays, and as a labeled reference for method development and metabolic studies.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for isotope-dilution quantitative assays.
  • High chemical purity for reliable analytical results.
  • Stable when stored under recommended low-temperature conditions.
  • Available in multiple small sizes for analytical workflows.