Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Hexanes
SCI_ED

Catalog No. S25352 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
ACS Grade
Lab Grade
Reagent Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25352 1 L Lab Grade
S25352A 4 L ACS Grade
S25351 500 mL Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25352
Add to Cart
Add to Cart

High-purity hexanes for laboratory experiments and chemistry education. Ideal for secondary and post-secondary science instruction.

  • Chemical Name: Hexanes (C6H14)
  • Grade: ACS Grade – suitable for educational and instructional lab use
  • Purity: Assay percent >95% w/v
  • Quantity: 4 liters
  • Colorless
  • Density: 0.659 g/mL
  • Formula Weight: 86.18
  • Use Case: Designed exclusively for educational use, Certificate of Analysis (C of A) not available
  • Packaging: Securely packaged for lab use in instructional settings

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Color Colorless
Density 0.659g/mL
Quantity 1 L
Packaging Lab Grade
Physical Form Liquid
Assay Percent Range >95% w/v
Linear Formula C6H14
Formula Weight 86.18
Chemical Name or Material Hexanes
ShelfLife 5 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.