Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, 95% n-hexane, HPLC Grade, J.T. Baker™

Catalog No. 02002996 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth PVC-coated Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-996 4 L Narrow Mouth PVC-coated Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-996 Supplier Avantor Supplier No. 930433
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity solvents for use in analytical and preparative separations

    For use in Liquid Chromatography (HPLC & UHPLC) & Spectrophotometry

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
IUPAC Name hexane
SMILES CCCCCC

Specifications

Density 0.663g/mL at 25°C (typical)
Quantity 4 L
Flash Point −23.00°C
Packaging Narrow Mouth PVC-coated Amber Glass Bottle
Specific Gravity 1L = 0.66kg
Boiling Point 68.7°C
Formula Weight 86.18
Percent Purity 95% n-hexane
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Hexanes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.