Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, 99% n-hexane, ULTRA RESI-ANALYZED™, J.T. Baker™

Catalog No. 02002993 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-993 4 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-993 Supplier Avantor Supplier No. N16808
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC); For Laboratory, Research, or Manufacturing Use

Specifications

CAS 110-54-3
Quantity 4 L
Flash Point −23.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.66kg
Molecular Formula C6H14
MDL Number MFCD02179311
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
PubChem CID 8058
ChEBI CHEBI:29021
Formula Weight 86.18
Percent Purity 99% n-hexane
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Hexanes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.