Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tritolyl phosphate, 63-85%, technical, residue other tolyl phosphate isomers

Catalog No. AC422315000
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422315000 500 g Glass bottle
AC422310250 25 g Glass bottle
AC422310025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC422315000 Supplier Thermo Scientific Chemicals Supplier No. 422315000
Only null left
Add to Cart

CAS: 1330-78-5 | C21H21O4P | 368.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-78-5
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.36
MDL Number MFCD02102116
Synonym Tricresyl phosphate

Specifications

Melting Point -30°C
Color White
Density 1.1600g/mL
Boiling Point 265°C (10.0 mmHg)
Flash Point 234°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (sum of isomers) (GC)
Packaging Glass bottle
Refractive Index 1.5550 to 1.557
Linear Formula (CH3C6H4O)3PO
Quantity 500 g
Specific Gravity 1.16
Merck Index 15, 9940
Solubility Information Solubility in water: insoluble
Formula Weight 368.36
Percent Purity 99%
Grade Technical
Physical Form Powder
Chemical Name or Material Tritolyl phosphate, tech., 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Collect spillage.
Avoid release to the en
EINECSNumber 215-548-8
RTECSNumber TD0175000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.