Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, 99%, pure, mixture of isomers

Catalog No. AC411540025
Change view
Click to view available options
Quantity:
2 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411540025 2.5 L Glass Bottle
AC411540010 1 L EcoSafPak™ Glass Bottle
AC411540100 10 L Plastic Drum
AC411540020 2 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC411540025 Supplier Thermo Scientific Chemicals Supplier No. 411540025
Add to Cart

CAS: 92112-69-1 | C6H14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanes
Melting Point -95.0°C
Name Note Mixture of Isomers
CAS 92112-69-1, 110-54-3
Color Colorless
Density 0.6590g/mL
Quantity 2.5 L
Flash Point −22°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3748 to 1.3810
Specific Gravity 0.659
Assay Percent Range 99%
Molecular Formula C6H14
Vapor Pressure Vapor Pressure: 160mbar at 20°C
Linear Formula CH3(CH2)4CH3
Viscosity 0.31 mPa.s (20°C)
MDL Number MFCD00009520
Beilstein 01, 142
Merck Index 14, 4694
Synonym Hex
Boiling Point 69.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Percent Purity 99%
Grade Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.