Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RSOH00101C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOH00101C 1 L
RSOH0010500 500 mL
RSOH00104C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RSOH00101C Supplier Ricca Chemical Company Supplier No. RSOH00101C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Max % 100
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.001%
Boiling Point 69°C
Percent Purity min. 98.5%
Grade ACS
Chemical Name or Material Hexanes
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.