Learn More
Hexanes, for HPLC, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 610070040
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCSpecifications
Hexanes | |
92112-69-1, 110-54-3 | |
C6H14 | |
MFCD00009520 | |
01, 142 | |
Hex | |
≥98.5% | |
0.6590g/mL | |
4 L | |
Pass Test | |
1.3748 to 1.3810 | |
0.02% max. |
98.5% | |
(1 cm cell vs water), 0.014 max. at 254nm, 0.08 max. at 220nm, 0.25 max. at 210nm, 1 max. at 195nm | |
CH3(CH2)4CH3 | |
1208 | |
14, 4694 | |
86.18 | |
HPLC | |
69.0°C | |
-22°C (71.4°F) | |
EcoSafPak™ | |
3ppm max. |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only