Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, for HPLC, contains ca. 95% n-Hexane

Catalog No. AC268360010
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268360010 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC268360010 Supplier Thermo Scientific Chemicals Supplier No. 268360010

missing translation for 'validMainframeMsg'

CAS: 92112-69-1 | C6H14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
MDL Number MFCD00009520
Synonym Hex

Specifications

Absorbance (1 cm cell vs HPLC−grade water), 0.005 max. at 250nm, 0.01 max. at 240nm, 0.03 max. at 230nm, 0.07 max. at 220nm, 0.25 max. at 210nm, 0.7 max. at 200nm
Density 0.6590g/mL
Quantity 1 L
Packaging Glass Bottle
Linear Formula CH3(CH2)4CH3
Vapor Pressure Vapor Pressure: 160mbar at 20°C
Beilstein 01, 142
Merck Index 14, 4694
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
Formula Weight 86.18
Grade HPLC
Chemical Name or Material Hexanes, (95% n-Hexane)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-777-6
RTECSNumber MN9275000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.