Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S031061 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031061 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING! : This product can expose you to Hexane, which is known to the State of California to cause birth defects or other reproductive harm.
Catalog No. S031061 Supplier Aldon Corporation Supplier No. IS17004
Add to Cart
Add to Cart
WARNING! : This product can expose you to Hexane, which is known to the State of California to cause birth defects or other reproductive harm.

Chemical Identifiers

CAS 110-54-3
Molecular Weight (g/mol) 86.18 g/mol

Specifications

Quantity 500 mL
Packaging Steel Conetop Can
Boiling Point 63 to 68°C
Percent Purity 70%
Grade Laboratory
Chemical Name or Material Hexanes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.