Learn More
Medchemexpress LLC Hexanoic acid, 3-oxo-, ethyl ester | 3249-68-1 | MFCD00009401 | 96.0% | 158.19 g/mol | C8H14O3 | 25 G
SDP
Catalog No. 5000303480
Encompass_Preferred
Ethyl butyrylacetate (ethyl 3-oxohexanoate) is an organic ester supplied as a laboratory reagent and synthetic intermediate. It is used in organic synthesis, flavor and fragrance chemistry, and as an analytical reference. Typical handling and storage guidance is provided by the manufacturer to preserve stability and purity.
  • High-purity reagent suitable for synthetic applications.
  • Identified by CAS 3249-68-1 for unambiguous chemical identity.
  • Available in multiple lab-scale pack sizes including 25 g.
  • Stable when stored under recommended conditions to maintain shelf life.
  • Useful as a building block and intermediate in multi-step organic syntheses.
Catalog No. 50-003-03480 Supplier Medchemexpress LLC Supplier No. 25GHYW015487
Ethyl butyrylacetate (ethyl 3-oxohexanoate) is an organic ester supplied as a laboratory reagent and synthetic intermediate. It is used in organic synthesis, flavor and fragrance chemistry, and as an analytical reference. Typical handling and storage guidance is provided by the manufacturer to preserve stability and purity.
  • High-purity reagent suitable for synthetic applications.
  • Identified by CAS 3249-68-1 for unambiguous chemical identity.
  • Available in multiple lab-scale pack sizes including 25 g.
  • Stable when stored under recommended conditions to maintain shelf life.
  • Useful as a building block and intermediate in multi-step organic syntheses.