Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexanoic acid, 3-oxo-, ethyl ester | 3249-68-1 | MFCD00009401 | 96.0% | 158.20 g·mol⁻1 | C8H14O3 | 50 G
SDP

Catalog No. 5000302547
Encompass_Preferred

Ethyl butyrylacetate (ethyl 3-oxohexanoate) is a ketoester used as a research reagent and synthetic intermediate in organic chemistry, offering a fruity aroma profile and reactivity useful for building complex molecules.

  • Useful synthetic building block for organic synthesis
  • Commonly used as a flavor and fragrance intermediate
  • Purity approximately 96.0% suitable for research applications
  • Molecular weight 158.20 g·mol⁻1; formula C8H14O3
  • Store at low temperature for extended stability (recommend -20°C)

Catalog No. 50-003-02547 Supplier Medchemexpress LLC Supplier No. HYW01548750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl butyrylacetate (ethyl 3-oxohexanoate) is a ketoester used as a research reagent and synthetic intermediate in organic chemistry, offering a fruity aroma profile and reactivity useful for building complex molecules.

  • Useful synthetic building block for organic synthesis
  • Commonly used as a flavor and fragrance intermediate
  • Purity approximately 96.0% suitable for research applications
  • Molecular weight 158.20 g·mol⁻1; formula C8H14O3
  • Store at low temperature for extended stability (recommend -20°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.