Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexanoic acid, 3-oxo-, ethyl ester | 3249-68-1 | MFCD00009401 | 96.0% | 158.20 g/mol | C8H14O3 | 10 G
SDP

Catalog No. 5000303754
Encompass_Preferred

Ethyl butyrylacetate (ethyl 3-oxohexanoate) is an organic ester used as a building block in organic synthesis and as a fragrance and flavor ingredient. It is supplied for laboratory use in multiple pack sizes and is provided with a certificate of analysis and a safety data sheet. Typical batch purity is approximately 96.0%.

  • CAS registry number 3249-68-1
  • Used as an intermediate in organic synthesis
  • Used as a fragrance and flavor component
  • Molecular formula C8H14O3 and molecular weight 158.20 g/mol
  • Available in laboratory pack sizes including 10 g
  • Certificate of analysis and safety data sheet available
  • Typical purity approximately 96.0%

Catalog No. 50-003-03754 Supplier Medchemexpress LLC Supplier No. HYW01548710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl butyrylacetate (ethyl 3-oxohexanoate) is an organic ester used as a building block in organic synthesis and as a fragrance and flavor ingredient. It is supplied for laboratory use in multiple pack sizes and is provided with a certificate of analysis and a safety data sheet. Typical batch purity is approximately 96.0%.

  • CAS registry number 3249-68-1
  • Used as an intermediate in organic synthesis
  • Used as a fragrance and flavor component
  • Molecular formula C8H14O3 and molecular weight 158.20 g/mol
  • Available in laboratory pack sizes including 10 g
  • Certificate of analysis and safety data sheet available
  • Typical purity approximately 96.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.