Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexanoic acid

Catalog No. 153745500G
Encompass
Change view
Click to view available options
Quantity:
2.5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-374-5500G 500 g
15-374-5100G 100 g
50-184-6783 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-374-5500G Supplier Sigma Aldrich Supplier No. 153745500G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexanoic acid

Specifications

CAS 142-62-1
Melting Point -4°C (lit.)
Density 0.927 g/mL (at 25°C (literature))
Boiling Point 202°C to 203°C (lit.)
Molecular Formula C6H12O2
Refractive Index n20/D 1.4161 (literature)
Linear Formula CH3(CH2)4 COOH
Quantity 500 g
MDL Number MFCD00004421
Synonym Acid C6; Caproic acid
Molecular Weight (g/mol) 116.16
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.