Learn More
Medchemexpress LLC Hexanoic acid, 6-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1-oxo-1H-isoindol-4-yl]amino]- | 2338824-30-7 | 96.4% | 373.40 | 5 MG
SDP
Catalog No. 5000394598
Encompass_Preferred
Lenalidomide-C5-acid is a Lenalidomide-based E3 ligase ligand-linker conjugate. It is used in the recruitment of CRBN protein and can be utilized to synthesize PROTACs. This product is for research use only and is not intended for human use.
  • E3 ligase ligand-linker conjugate based on Lenalidomide.
  • Used in the recruitment of CRBN protein.
  • Can be used to synthesize PROTACs.
Catalog No. 50-003-94598 Supplier Medchemexpress LLC Supplier No. HY1602415MG
Lenalidomide-C5-acid is a Lenalidomide-based E3 ligase ligand-linker conjugate. It is used in the recruitment of CRBN protein and can be utilized to synthesize PROTACs. This product is for research use only and is not intended for human use.
  • E3 ligase ligand-linker conjugate based on Lenalidomide.
  • Used in the recruitment of CRBN protein.
  • Can be used to synthesize PROTACs.