Learn More
Medchemexpress LLC Hexanoic acid, 6-azido-, 2,5-dioxo-1-pyrrolidinyl ester | 866363-70-4 | MFCD25968077 | ≥98.0% | 254.24 g/mol | C10H14N4O4 | 1 G
SDP
Catalog No. 5000202953
Encompass_Preferred
N3-C5-NHS ester is an azide-functionalized N-hydroxysuccinimide (NHS) ester used as a noncleavable ADC linker and a click-chemistry reagent for bioconjugation. It enables coupling to primary amines via NHS chemistry and to alkynes via azide-alkyne cycloaddition reactions, and is supplied as a colorless to light yellow liquid with high purity.
  • Noncleavable ADC linker suitable for antibody-drug conjugation
  • Enables conjugation to primary amines via NHS ester chemistry
  • Compatible with copper-catalyzed and strain-promoted azide-alkyne click reactions
  • Azide functionality provides orthogonal bioconjugation options
  • Supplied as a 1 g quantity for laboratory use
Catalog No. 50-002-02953 Supplier Medchemexpress LLC Supplier No. HY1265231G
N3-C5-NHS ester is an azide-functionalized N-hydroxysuccinimide (NHS) ester used as a noncleavable ADC linker and a click-chemistry reagent for bioconjugation. It enables coupling to primary amines via NHS chemistry and to alkynes via azide-alkyne cycloaddition reactions, and is supplied as a colorless to light yellow liquid with high purity.
  • Noncleavable ADC linker suitable for antibody-drug conjugation
  • Enables conjugation to primary amines via NHS ester chemistry
  • Compatible with copper-catalyzed and strain-promoted azide-alkyne click reactions
  • Azide functionality provides orthogonal bioconjugation options
  • Supplied as a 1 g quantity for laboratory use