Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexanoic acid, 6-azido-, 2,5-dioxo-1-pyrrolidinyl ester | 866363-70-4 | MFCD25968077 | 98.0% | 254.24 g/mol | C10H14N4O4 | 250 MG
SDP

Catalog No. 5000203457
Encompass_Preferred

N3-C5-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker and click chemistry reagent that contains an azide group and an N-hydroxysuccinimide (NHS) ester for efficient amine coupling. It is used to introduce azide handles for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and to activate carboxyl groups for conjugation to biomolecules.

  • Noncleavable ADC linker suitable for bioconjugation
  • Contains azide group for click chemistry (CuAAC)
  • NHS ester functional group for coupling to primary amines
  • Purity ≥98.0%
  • Supplied as a colorless to light yellow liquid; soluble in DMSO
  • Molecular weight 254.24 g/mol; formula C10H14N4O4
  • Storage: pure form -20°C; in solvent short-term at -20°C to -80°C

Catalog No. 50-002-03457 Supplier Medchemexpress LLC Supplier No. HY126523250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N3-C5-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker and click chemistry reagent that contains an azide group and an N-hydroxysuccinimide (NHS) ester for efficient amine coupling. It is used to introduce azide handles for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and to activate carboxyl groups for conjugation to biomolecules.

  • Noncleavable ADC linker suitable for bioconjugation
  • Contains azide group for click chemistry (CuAAC)
  • NHS ester functional group for coupling to primary amines
  • Purity ≥98.0%
  • Supplied as a colorless to light yellow liquid; soluble in DMSO
  • Molecular weight 254.24 g/mol; formula C10H14N4O4
  • Storage: pure form -20°C; in solvent short-term at -20°C to -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.