Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexanoic acid 98 fCC fG5KG
Catalog No. 502897502
Encompass
Hexanoic acid is a precursor of ethyl caproate the key flavor constituent of Chinese strong-flavored liquor (CSfL).
Catalog No. 50-289-7502 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2559045KGK
Hexanoic acid is a precursor of ethyl caproate the key flavor constituent of Chinese strong-flavored liquor (CSfL).