Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanoic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018935 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8935 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8935 Supplier MilliporeSigma Supelco Supplier No. 215295ML
Add to Cart
Add to Cart

Hexanoic acid is a six-carbon saturated fatty acid found naturally in oils and animal fats as well as produced by certain anaerobic bacteria. It has been widely used in industrial applications such as perfumes, medicine, food additives, lubricating grease, tobacco flavor, rubber, and dyes.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 142-62-1
Vapor Pressure 0.18 mmHg (20°C)
Vapor Density 4 (vs. air)
Density 0.927 g/mL (at 25°C)
Flash Point 102°C
Physical Form Neat
Linear Formula CH3(CH2)4 COOH
MDL Number MFCD00004421
Melting Point -4°C
Boiling Point 202°C to 203°C
Synonym Acid C6; Caproic acid
Type Analytical Standard
Quantity 5 mL
Formula Weight 116.16
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.