Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexanoyl chloride

Catalog No. 29465925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-465-925ML 25 mL
29-465-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-465-925ML Supplier Sigma Aldrich Supplier No. 29465925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Hexanoyl chloride has been used in:
  • Total synthesis of (+/-)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one
  • Asymmetric synthesis of 14-methyl-1-octadecene, the sex pheromone of the peach leafminer moth
  • Preparation of hexanoyl-coated nanofibers dispersible in several organic solvents
  • Total synthesis of natural isocarbostyril ruprechstyril (3-n-pentyl-6-methoxy-8-hydroxy-1(2h)-isoquinolinone), isolated from ruprechtia tangarana
  • 5-chloro-8-hydroxy-6-methoxy-3-pentylisocoumarin, the 5-chloro analog of naturally occurring 7-chloro-8-hydroxy-6-methoxy-3-pentylisocoumarin, isolated from tessmannia densiflora

Specifications

CAS 142-61-0
Density 0.963 g/mL (at 25°C (literature))
Molecular Formula C6H11ClO
Linear Formula CH3(CH2)4 COCl
MDL Number MFCD00000760
Boiling Point 150°C to 153°C (lit.)
Synonym Caproyl chloride
Molecular Weight (g/mol) 134.6
Quantity 25 mL
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.