Learn More
Abcam Hexanoyl-Lysine BSA Conjugate, 50UG
Catalog No. 502704220
Encompass_Preferred
Hexanoyl-Lysine BSA Conjugate is a Cow Full Length protein, expressed in Synthetic, with >98% purity and suitable for SDS-PAGE, WB, ELISA.
Protein/ Peptide/ Enzyme Synonyms: Albumin, BSA, ALB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4220 Supplier Abcam Supplier No. AB24591850UG
Hexanoyl-Lysine BSA Conjugate is a Cow Full Length protein, expressed in Synthetic, with >98% purity and suitable for SDS-PAGE, WB, ELISA.
Protein/ Peptide/ Enzyme Synonyms: Albumin, BSA, ALB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement