Learn More
Medchemexpress LLC (±)-Hexanoylcarnitine chloride | 6920-35-0 | MFCD00151472 | 98.0% | 295.80 | C13H26ClNO4 | 25 MG
SDP
Catalog No. 5000212050
Encompass_Preferred
Racemic hexanoylcarnitine chloride is a racemic acylcarnitine used as an analytical standard and research reagent in metabolomics and biomarker studies. It is a medium-chain fatty-acid metabolite involved in fatty acid oxidation and is applied in assays for skeletal muscle toxicity and metabolic profiling.
  • Used as an analytical standard for metabolomics and quantitative assays.
  • Racemic (±) acylcarnitine suitable for comparative studies.
  • Acts as a medium-chain fatty-acid metabolite involved in fatty acid oxidation.
  • Serves as a detectable biomarker for skeletal muscle toxicity and disease association studies.
  • Stable solid formulation with defined storage recommendations.
Catalog No. 50-002-12050 Supplier Medchemexpress LLC Supplier No. HY10097825MG
Racemic hexanoylcarnitine chloride is a racemic acylcarnitine used as an analytical standard and research reagent in metabolomics and biomarker studies. It is a medium-chain fatty-acid metabolite involved in fatty acid oxidation and is applied in assays for skeletal muscle toxicity and metabolic profiling.
  • Used as an analytical standard for metabolomics and quantitative assays.
  • Racemic (±) acylcarnitine suitable for comparative studies.
  • Acts as a medium-chain fatty-acid metabolite involved in fatty acid oxidation.
  • Serves as a detectable biomarker for skeletal muscle toxicity and disease association studies.
  • Stable solid formulation with defined storage recommendations.