Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexanoylcarnitine chloride | 6920-35-0 | MFCD00151472 | 98.0% | 295.80 g/mol | C13H26ClNO4 | 10 MG
SDP

Catalog No. 5000211435
Encompass_Preferred

(±)-Hexanoylcarnitine chloride is a racemic acylcarnitine compound used as an analytical standard and biomarker in metabolomics and biochemical research. Supplied as a high-purity solid, it is intended for analytical assays and reference applications.

  • High purity 98.0%.
  • Solid, white to off-white appearance.
  • Molecular weight 295.80 g/mol.
  • Available in milligram-scale pack sizes for analytical use.
  • Store at 4°C in a sealed container to protect from moisture.
  • Suitable as a biomarker reference in metabolomics studies.

Catalog No. 50-002-11435 Supplier Medchemexpress LLC Supplier No. HY10097810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Hexanoylcarnitine chloride is a racemic acylcarnitine compound used as an analytical standard and biomarker in metabolomics and biochemical research. Supplied as a high-purity solid, it is intended for analytical assays and reference applications.

  • High purity 98.0%.
  • Solid, white to off-white appearance.
  • Molecular weight 295.80 g/mol.
  • Available in milligram-scale pack sizes for analytical use.
  • Store at 4°C in a sealed container to protect from moisture.
  • Suitable as a biomarker reference in metabolomics studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.