Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexanoylglycine-d2 | 1256842-52-0 | 175.22 | 25 MG
SDP

Catalog No. 5000439995
Encompass_Preferred

Hexanoylglycine-d2 is a chemical product identified for laboratory use and substance manufacturing. It is supplied as a solid substance.

  • Chemical name: Hexanoylglycine-d2
  • CAS number: 1256842-52-0
  • Molecular weight: 175.22
  • Appearance: Solid
  • Recommended storage temperature: powder: -20°C for 3 years; 4°C for 2 years. In solvent: -80°C for 6 months.

Catalog No. 50-004-39995 Supplier Medchemexpress LLC Supplier No. 25MGHY113150S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexanoylglycine-d2 is a chemical product identified for laboratory use and substance manufacturing. It is supplied as a solid substance.

  • Chemical name: Hexanoylglycine-d2
  • CAS number: 1256842-52-0
  • Molecular weight: 175.22
  • Appearance: Solid
  • Recommended storage temperature: powder: -20°C for 3 years; 4°C for 2 years. In solvent: -80°C for 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.