Learn More
Strem, An Ascensus Company CAS# 28407-51-4. 250mg. Hexarhodium hexadecacarbonyl, min. 98%. MFCD00011206
SDP
Supplier:  Strem, An Ascensus Company 450820250MG
Encompass
CAS# 28407-51-4. 250mg. Hexarhodium hexadecacarbonyl, min. 98%. MFCD00011206. Molecular Weight: 1065.61. Molecular Formula: Rh6(CO)16. Color/form: black xtl. Strem# 45-0820. http://www.strem.com/catalog/v/45-0820/
Catalog No. 50-901-12196