Learn More
MSC HexArmor Arm Guard, 8 in, ANSI Cut A8, 1 ea
Catalog No. 5000782783
Encompass
This 8 in arm guard provides industrial puncture protection for the forearm, featuring a spandex wrist insert with a thumbhole and snap fasteners. It is engineered to prevent snags, punctures, abrasions, or cuts to the forearm, utilizing advanced SuperFabric® material.
  • Highest needlestick resistance due to SuperFabric® brand material layering
  • Tested using actual 25 gauge needles
  • High level of cut-resistance on enhanced areas
  • Lightweight and cooler than knit or leather alternatives
  • Designed to stay up without falling down
  • Launderable
Catalog No. 50-007-82783 Supplier MSC Supplier No. 91430652
This 8 in arm guard provides industrial puncture protection for the forearm, featuring a spandex wrist insert with a thumbhole and snap fasteners. It is engineered to prevent snags, punctures, abrasions, or cuts to the forearm, utilizing advanced SuperFabric® material.
  • Highest needlestick resistance due to SuperFabric® brand material layering
  • Tested using actual 25 gauge needles
  • High level of cut-resistance on enhanced areas
  • Lightweight and cooler than knit or leather alternatives
  • Designed to stay up without falling down
  • Launderable