These hi-vis gloves are designed to protect against prevalent worksite hazards, featuring SuperFabric® brand material on the palm and advanced impact-resistant guards on the back of the hand.
- SuperFabric® material provides cut, puncture, and abrasion resistance in the palm.
- Hi-vis back-of-hand impact guards protect against smash and pinch hazards.
- Level 2 contact heat rated.
- Mud Grip® palm with synthetic leather and PVC dots offers reliable grip and increased abrasion resistance.
- Reinforced interior seams with double-stitched core-spun thread for durability.
- Hi-vis color scheme and reflective piping increase visual awareness.
- Elastic cuff with hook and loop closure and pull tab for secure fit.
- Form-fitting materials for increased dexterity and comfort.
- Launderable and vend-packed.