HexArmor Chrome series gloves provide advanced cut, puncture, and abrasion resistance, utilizing SuperFabric material. These gloves feature high-visibility back-of-hand impact guards and a Mud Grip palm for enhanced grip in dry or light oil conditions.
- SuperFabric material provides industry-leading cut, puncture, and abrasion resistance on the palm.
- High-visibility back-of-hand impact guards protect against smash and pinch hazards.
- Mud Grip palm with synthetic leather and PVC dots for reliable grip in dry or light oil.
- Interior seams reinforced with double-stitched core-spun thread.
- High-visibility color scheme and reflective tape for increased visual awareness.
- Elastic cuff with secure hook and loop closure.
- Pull tab for easy donning and doffing.
- Form-fitting materials for increased dexterity and comfort.
- Launderable.