Learn More
MSC HexArmor Cut-Resistant Sleeve, Aramid, Small, ANSI Cut A6, 1 ea
Catalog No. 5000768942
Encompass
This cut-resistant sleeve is designed to provide high-level protection against cuts. Constructed from Aramid material, it features a small size with a gray and green color scheme, offering both comfort and a secure fit for various applications.
  • Cut-resistant sleeves
  • Aramid material
  • Small sleeve size
  • Numeric size: 7
  • Gray and green color
  • Knit wrist closure
  • Overall length: 18.00 in.
  • Thumb hole included
  • Knit seam style
  • Cut-resistant properties
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA puncture resistance level 0
  • ANSI/ISEA cut resistance level A6
  • Flame resistant
  • Made in China
Catalog No. 50-007-68942 Supplier MSC Supplier No. 80802499
This cut-resistant sleeve is designed to provide high-level protection against cuts. Constructed from Aramid material, it features a small size with a gray and green color scheme, offering both comfort and a secure fit for various applications.
  • Cut-resistant sleeves
  • Aramid material
  • Small sleeve size
  • Numeric size: 7
  • Gray and green color
  • Knit wrist closure
  • Overall length: 18.00 in.
  • Thumb hole included
  • Knit seam style
  • Cut-resistant properties
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA puncture resistance level 0
  • ANSI/ISEA cut resistance level A6
  • Flame resistant
  • Made in China