Learn More
MSC HexArmor Cut-Resistant Work Glove, Cut A6, X-Large, 1 Pair
Catalog No. 5000685090
Encompass
The Chrome Series 4018 glove is a premier maintenance and repair glove, featuring SuperFabric® brand material on the palm and knuckle patch for superior protection, combined with a synthetic leather grip and touchscreen capabilities for versatile use.
  • SuperFabric® brand material on the palm and knuckle patch provides industry-leading cut, puncture, and abrasion resistance
  • Synthetic leather palm offers reliable grip in dry or light oil situations and increases abrasion resistance
  • Touchscreen capabilities on the thumb and index fingertip
  • Interior seams are reinforced with a double-stitched core-spun thread for enhanced durability
  • Neoprene cuff with hook and loop wrist closure ensures a comfortable, secure fit
  • Pull tab for easy don and doff
  • Launderable
  • Vend-packed
Catalog No. 50-006-85090 Supplier MSC Supplier No. 82149758
The Chrome Series 4018 glove is a premier maintenance and repair glove, featuring SuperFabric® brand material on the palm and knuckle patch for superior protection, combined with a synthetic leather grip and touchscreen capabilities for versatile use.
  • SuperFabric® brand material on the palm and knuckle patch provides industry-leading cut, puncture, and abrasion resistance
  • Synthetic leather palm offers reliable grip in dry or light oil situations and increases abrasion resistance
  • Touchscreen capabilities on the thumb and index fingertip
  • Interior seams are reinforced with a double-stitched core-spun thread for enhanced durability
  • Neoprene cuff with hook and loop wrist closure ensures a comfortable, secure fit
  • Pull tab for easy don and doff
  • Launderable
  • Vend-packed