Learn More
MSC HexArmor Face Shield Window, Clear, 4 in, Anti-Fog/Scratch-Resistant, 1 ea
Catalog No. 5000787444
Encompass
This clear replacement window is engineered for HexArmor face shields, offering reliable protection for various applications. It features advanced coatings for enhanced visibility and durability, meeting critical safety standards for chemical splash protection.
  • Compatible with HexArmor Ceros Safety Helmets
  • Features permanent TruShield® anti-fog coating on the inside
  • Includes scratch-resistant hard coat on the outside
  • Meets ANSI Z87.1-2015, Z87+, CE EN166, and CSA Z94.3 standards
  • Provides D3 liquid splash protection
  • Offers 100% UV protection
  • Easy and quick attachment mechanism
  • Designed for chemical splash applications
Catalog No. 50-007-87444 Supplier MSC Supplier No. 18752204
This clear replacement window is engineered for HexArmor face shields, offering reliable protection for various applications. It features advanced coatings for enhanced visibility and durability, meeting critical safety standards for chemical splash protection.
  • Compatible with HexArmor Ceros Safety Helmets
  • Features permanent TruShield® anti-fog coating on the inside
  • Includes scratch-resistant hard coat on the outside
  • Meets ANSI Z87.1-2015, Z87+, CE EN166, and CSA Z94.3 standards
  • Provides D3 liquid splash protection
  • Offers 100% UV protection
  • Easy and quick attachment mechanism
  • Designed for chemical splash applications