Learn More
MSC HexArmor Glove, SuperFabric, ANSI Cut A7, 19 in, 1 ea
Catalog No. 5000787638
Encompass
This protective glove features an extended sleeve constructed with SuperFabric® material, offering exceptional resistance to cuts, abrasions, and punctures for demanding applications.
  • Constructed from SuperFabric
  • Sleeve size is 3X-Large
  • Glove size is 12
  • Black color
  • Hook and loop closure
  • Overall length is 19.00 in
  • Includes thumb hole
  • Cut-resistant
  • ANSI/ISEA abrasion resistance level 6
  • ANSI/ISEA puncture resistance level 3
  • ANSI/ISEA cut resistance level A7
  • Made in Indonesia
Catalog No. 50-007-87638 Supplier MSC Supplier No. 59346205
This protective glove features an extended sleeve constructed with SuperFabric® material, offering exceptional resistance to cuts, abrasions, and punctures for demanding applications.
  • Constructed from SuperFabric
  • Sleeve size is 3X-Large
  • Glove size is 12
  • Black color
  • Hook and loop closure
  • Overall length is 19.00 in
  • Includes thumb hole
  • Cut-resistant
  • ANSI/ISEA abrasion resistance level 6
  • ANSI/ISEA puncture resistance level 3
  • ANSI/ISEA cut resistance level A7
  • Made in Indonesia