The Helix 2059 is a high-dexterity winter work glove designed to provide protection from cold, featuring 360° cut-resistance, a three-quarter sandy nitrile coating, and touchscreen capabilities. Its interior fleece lining ensures warmth while maintaining dexterity in cold environments.
- Eighteen-gauge acrylic and fiberglass blend shell provides 360° cut resistance.
- Flexible sandy nitrile three-quarter coating offers superior grip and abrasion resistance.
- Fleece lining for warmth and insulation.
- Hi-vis color enhances visibility and compliance.
- Black palm coating hides dirt and soil.
- Touchscreen capabilities.
- Elastic knit wrist helps prevent dirt and debris from entering the glove.
- Seamless construction for enhanced comfort and breathability.
- Launderable for extended life.