Learn More
MSC HexArmor Gloves, Cut A4, Acrylic and Nitrile, X-Large, 1 Pair
Catalog No. 5000674401
Encompass
These cut, puncture, and abrasion-resistant gloves are constructed with an Acrylic primary material and feature a Nitrile coating on the palm and fingertips for a textured grip.
  • Cut, puncture and abrasion-resistant
  • Nitrile coating on palm and fingertips
  • Textured grip surface
  • Acrylic primary material
  • Fiberglass knit and acrylic back material
  • Fiberglass and acrylic lining material
  • Black with high-visibility yellow color
  • Gauge: 18
  • Size: X-Large
  • Overall length: 11 in.
  • Cuff style: Knit
  • Cuff color: Hi-visibility
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA puncture resistance level 2
  • ANSI/ISEA abrasion resistance level 4
  • EN 388 rating 3X42D
Catalog No. 50-006-74401 Supplier MSC Supplier No. 73987570
These cut, puncture, and abrasion-resistant gloves are constructed with an Acrylic primary material and feature a Nitrile coating on the palm and fingertips for a textured grip.
  • Cut, puncture and abrasion-resistant
  • Nitrile coating on palm and fingertips
  • Textured grip surface
  • Acrylic primary material
  • Fiberglass knit and acrylic back material
  • Fiberglass and acrylic lining material
  • Black with high-visibility yellow color
  • Gauge: 18
  • Size: X-Large
  • Overall length: 11 in.
  • Cuff style: Knit
  • Cuff color: Hi-visibility
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA puncture resistance level 2
  • ANSI/ISEA abrasion resistance level 4
  • EN 388 rating 3X42D