Learn More
MSC HexArmor Gloves, Cut A5, Puncture 4, 2X-Large, 1 Pair
Catalog No. 5000796331
Encompass
HexArmor Helix Core series 3012 impact gloves provide high levels of cut, puncture, and abrasion resistance, ideal for demanding manufacturing. They feature back-of-hand protection, textured grip, and touchscreen capabilities, ensuring durability, protection, and comfort for precision tasks.
  • Features 15-gauge HPPE/steel/fiberglass/polyester blend shell with 360° cut resistance
  • Polyurethane palm coating offers superior grip and abrasion resistance
  • Sewn-on back-of-hand IR-X® Impact Exoskeleton™ with high-flex design
  • Reinforced thumb saddle; enhanced touchscreen capabilities
  • Elastic knit wrist prevents dirt and debris entry
  • Seamless construction for comfort and breathability
  • Launderable for extended glove life
  • ANSI A5 cut, ANSI 4 puncture, and ANSI 5 abrasion resistance
Catalog No. 50-007-96331 Supplier MSC Supplier No. 26236216
HexArmor Helix Core series 3012 impact gloves provide high levels of cut, puncture, and abrasion resistance, ideal for demanding manufacturing. They feature back-of-hand protection, textured grip, and touchscreen capabilities, ensuring durability, protection, and comfort for precision tasks.
  • Features 15-gauge HPPE/steel/fiberglass/polyester blend shell with 360° cut resistance
  • Polyurethane palm coating offers superior grip and abrasion resistance
  • Sewn-on back-of-hand IR-X® Impact Exoskeleton™ with high-flex design
  • Reinforced thumb saddle; enhanced touchscreen capabilities
  • Elastic knit wrist prevents dirt and debris entry
  • Seamless construction for comfort and breathability
  • Launderable for extended glove life
  • ANSI A5 cut, ANSI 4 puncture, and ANSI 5 abrasion resistance