The HexArmor Chrome Series® 4026 gloves are engineered for superior protection against common worksite hazards. They feature SuperFabric® brand material on the palm for industry-leading cut, puncture, and abrasion resistance, combined with advanced, low-profile impact-resistant guards on the back of the hand.
- SuperFabric® palm provides A8 cut, puncture, and abrasion resistance
- Low-profile impact guards on back-of-hand protect knuckles and fingers
- Level 2 contact heat rated
- Mud Grip® palm with synthetic leather + PVC dots for reliable grip in wet conditions
- Reinforced interior seams with double-stitched thread for durability
- Hi-vis color scheme and reflective piping enhance visual awareness
- Elastic cuff with hook and loop closure and pull tab for secure fit
- Form-fitting materials increase dexterity and comfort
- Launderable