Learn More
MSC HexArmor Helix 2059 Gloves, Cut A4, Medium, 1 Pair
Catalog No. 5000796939
Encompass
These cut, puncture, and abrasion-resistant gloves feature an acrylic, nitrile-coated palm and fingertips with a textured grip, designed for demanding applications.
  • Knit style with nitrile coating on palm and fingertips
  • Cut, puncture, and abrasion-resistant for enhanced protection
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 2
  • EN 388 rating 3X42D for comprehensive protection
  • Constructed with 18 gauge fiberglass knit and acrylic materials
  • Measures 11 in overall length
Catalog No. 50-007-96939 Supplier MSC Supplier No. 73986994
These cut, puncture, and abrasion-resistant gloves feature an acrylic, nitrile-coated palm and fingertips with a textured grip, designed for demanding applications.
  • Knit style with nitrile coating on palm and fingertips
  • Cut, puncture, and abrasion-resistant for enhanced protection
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 2
  • EN 388 rating 3X42D for comprehensive protection
  • Constructed with 18 gauge fiberglass knit and acrylic materials
  • Measures 11 in overall length