Learn More
MSC HexArmor Helix Gloves, Cut A2/Puncture 3/Abrasion 5, Medium, 1 Pair
Catalog No. 5000767208
Encompass
These gloves offer cut, puncture, and abrasion resistance, designed for demanding applications requiring hand protection. They feature a durable HPPE blend construction with a nitrile coating on the palm and fingertips.
  • Knit style provides comfort and dexterity
  • Nitrile coating ensures enhanced grip and durability
  • ANSI/ISEA Cut Resistance Level A2 for moderate cut hazards
  • ANSI/ISEA Puncture Resistance Level 3 for significant puncture protection
  • ANSI/ISEA Abrasion Resistance Level 5 for high abrasion resistance
  • HPPE blend lining for robust protection
  • Palm and fingertips coating for targeted protection
  • Provides 21 gauge construction for a balance of protection and flexibility
  • Blue with high-visibility yellow elements
Catalog No. 50-007-67208 Supplier MSC Supplier No. 29807757
These gloves offer cut, puncture, and abrasion resistance, designed for demanding applications requiring hand protection. They feature a durable HPPE blend construction with a nitrile coating on the palm and fingertips.
  • Knit style provides comfort and dexterity
  • Nitrile coating ensures enhanced grip and durability
  • ANSI/ISEA Cut Resistance Level A2 for moderate cut hazards
  • ANSI/ISEA Puncture Resistance Level 3 for significant puncture protection
  • ANSI/ISEA Abrasion Resistance Level 5 for high abrasion resistance
  • HPPE blend lining for robust protection
  • Palm and fingertips coating for targeted protection
  • Provides 21 gauge construction for a balance of protection and flexibility
  • Blue with high-visibility yellow elements