Learn More
MSC HexArmor Helix Gloves, Cut A5/P4/A5, X-Small, 1 Pair
Catalog No. 5000770320
Encompass
These gloves are designed for cut, puncture, and abrasion resistance, featuring an HPPE blend and a polyurethane coating on the palm and fingertips for enhanced protection.
  • Knit style for comfort
  • Polyurethane coating material
  • Coating covers palm and fingertips
  • Cut, puncture, and abrasion-resistant glove type
  • ANSI/ISEA cut resistance level A5
  • Primary material HPPE blend
  • ANSI/ISEA abrasion resistance level 5
  • Blue and high-visibility yellow color
  • X-small glove size (numeric size 6)
  • Helix series
  • ANSI/ISEA puncture resistance level 4
  • Black coating color
  • Features a 15 gauge knit
  • EN 388 rating 4X43DP
  • TPR back material
  • HPPE blend lining material
  • Supplied as a pair
  • Knit cuff style
  • Unisex design
Catalog No. 50-007-70320 Supplier MSC Supplier No. 16109456
These gloves are designed for cut, puncture, and abrasion resistance, featuring an HPPE blend and a polyurethane coating on the palm and fingertips for enhanced protection.
  • Knit style for comfort
  • Polyurethane coating material
  • Coating covers palm and fingertips
  • Cut, puncture, and abrasion-resistant glove type
  • ANSI/ISEA cut resistance level A5
  • Primary material HPPE blend
  • ANSI/ISEA abrasion resistance level 5
  • Blue and high-visibility yellow color
  • X-small glove size (numeric size 6)
  • Helix series
  • ANSI/ISEA puncture resistance level 4
  • Black coating color
  • Features a 15 gauge knit
  • EN 388 rating 4X43DP
  • TPR back material
  • HPPE blend lining material
  • Supplied as a pair
  • Knit cuff style
  • Unisex design