Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ Hercules™ NSR 3041 Gloves

Catalog No. 19160603 Shop All Performance Fabrics, Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-160-603 X-Large
19-130-5445 Medium
19-130-5446 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-160-603 Supplier HexArmor™ Supplier No. 3041XL 10
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ANSI/ISEA Cut Level A9 gloves feature full coverage design and pre-curved shape for comfort and protection.

  • SuperFabric™ brand material offers highest needlestick-resistance available
  • 360° degree cut protection - ANSI/ISEA Cut Level A9, Needlestick Level 5
  • Full coverage design and pre-curved shape for comfort and protection
  • Specialized silicone gripping surface on palm
  • Launderable

Specifications

Coating Material Silicone
Color Black
Cut Resistance Rating ANSI A9
Material SuperFabric
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388:4X12F
For Use With (Application) Fine-toothed animal handling, wastewater and sewage treatment
Model 3041-XL
Series Hercules™ NSR 3041
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.