Learn More
MSC HexArmor LT300 Safety Goggles, OTG, TruShield, ANSI Z87.1.2015, 1 ea
Catalog No. 5000686563
Encompass
The LT300 safety goggles offer durable protection against liquids, dirt, and debris with a soft seal and indirect ventilation. They fit over most prescription glasses and provide a wide field of vision.
  • Fits over most prescription glasses
  • Panoramic lens for wide field of vision
  • LTX Lens Technology for premium optical clarity
  • Offers 100% UV protection
  • Compliant with ANSI Z87.1.2015 and CE EN166
  • Permanent anti-fog and scratch-resistant TruShield coatings
Catalog No. 50-006-86563 Supplier MSC Supplier No. 70322649
The LT300 safety goggles offer durable protection against liquids, dirt, and debris with a soft seal and indirect ventilation. They fit over most prescription glasses and provide a wide field of vision.
  • Fits over most prescription glasses
  • Panoramic lens for wide field of vision
  • LTX Lens Technology for premium optical clarity
  • Offers 100% UV protection
  • Compliant with ANSI Z87.1.2015 and CE EN166
  • Permanent anti-fog and scratch-resistant TruShield coatings