Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ Rig Lizard™ 2021 Impact Protection Gloves

Catalog No. 19163520 Shop All Performance Fabrics, Inc Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-163-520 Small
19-163-521 Medium
19-163-522 Large
19-163-523 X-Large
19-163-524 2X-Large
19-163-525 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-163-520 Supplier HexArmor™ Supplier No. 2021S
This item is not returnable. View return policy

Gloves feature IR-X™ Impact Exoskeleton™ on the back of the hand to protect from impacts and smashing. Palms are constructed from TP-X™ with a reinforced stitching pattern for dexterity, and grip abilities.

  • IR-X™ Impact Exoskeleton™ with high-flex design keeps hands safe from smash & impact hazards
  • Reinforced index finger and thumb crotch for increased longevity
  • SlipFit™ cuff adds comfort and convenience 
  • Durable TP-X™ palm with reinforced stitching pattern
  • Hi-vis color scheme for increased visual awareness on the job site
  • Launderable

Specifications

Certifications/Compliance ANSI/ISEA 138 Level 1 Impact Protection
Cuff Material SlipFit™
Color Hi-vis Lime Green, Black
Cut Resistance Rating ANSI A3
Hand Specific
Material IR-X Impact Exoskeleton
Product Type Mechanic's Gloves
Size Small
For Use With (Application) General Work, Extrication, Oil/Gas Operations, Machine Repair, and Mechanics
Puncture Rating ASTM F2878 LEVEL 4
Series Rig Lizard™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.