Learn More
MSC HexArmor Safety Glasses, Anti-Fog, Scratch-Resistant, Gray Polycarbonate, Full-Framed, 1 ea
Catalog No. 5000788960
Encompass
These safety glasses feature anti-fog and scratch-resistant gray polycarbonate lenses in a full-framed design, meeting ANSI Z87.1.2015 standards. They are designed for reliable eye protection in various work environments.
  • Tinted glasses style
  • Anti-fog and scratch-resistant lens coating
  • Full-framed and wrap around design
  • Gray polycarbonate lenses
  • Yellow and charcoal frame with gray temples
  • Universal size
  • Adjustable and flexible temples
  • UV protection
  • Meets ANSI Z87.1.2015 standards
  • Includes gasket for enhanced protection
  • TPE frame material
  • Anti-fog coatings improve visibility under hot and humid conditions
Catalog No. 50-007-88960 Supplier MSC Supplier No. 19715432
These safety glasses feature anti-fog and scratch-resistant gray polycarbonate lenses in a full-framed design, meeting ANSI Z87.1.2015 standards. They are designed for reliable eye protection in various work environments.
  • Tinted glasses style
  • Anti-fog and scratch-resistant lens coating
  • Full-framed and wrap around design
  • Gray polycarbonate lenses
  • Yellow and charcoal frame with gray temples
  • Universal size
  • Adjustable and flexible temples
  • UV protection
  • Meets ANSI Z87.1.2015 standards
  • Includes gasket for enhanced protection
  • TPE frame material
  • Anti-fog coatings improve visibility under hot and humid conditions