Learn More
MSC HexArmor Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, Half-Framed, 1 ea
Catalog No. 5000789050
Encompass
These safety glasses feature anti-fog and scratch-resistant green polycarbonate lenses with a half-framed design. They meet standards ANSI Z87+, CSA Z94.3, and EN 166, providing reliable eye protection for various applications.
  • Anti-fog and scratch-resistant lens coating
  • Half-framed, wrap-around design
  • Green polycarbonate lenses
  • Adjustable temples
  • UV protection
  • Includes gasket
  • TPE frame material
  • Contoured temple type
  • Meets ANSI Z87+, CSA Z94.3, and EN 166 standards
Catalog No. 50-007-89050 Supplier MSC Supplier No. 51321651
These safety glasses feature anti-fog and scratch-resistant green polycarbonate lenses with a half-framed design. They meet standards ANSI Z87+, CSA Z94.3, and EN 166, providing reliable eye protection for various applications.
  • Anti-fog and scratch-resistant lens coating
  • Half-framed, wrap-around design
  • Green polycarbonate lenses
  • Adjustable temples
  • UV protection
  • Includes gasket
  • TPE frame material
  • Contoured temple type
  • Meets ANSI Z87+, CSA Z94.3, and EN 166 standards